siRNA Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256500.2 | XP_006256562.1 | 13 | 2589 | ||
XM_008773011.1 | XP_008771233.1 | 15 | 2778 | |||
XM_008773012.1 | XP_008771234.1 | 15 | 2778 | |||
XM_008773013.1 | XP_008771235.1 | 14 | 2631 | |||
XM_008773014.1 | XP_008771236.1 | 13 | 2589 | |||
XM_008773015.1 | XP_008771237.1 | 16 | 2921 | |||
XM_008773016.1 | XP_008771238.1 | 15 | 2879 |

More Information
Other Species Targeted

