siRNA Details
| Gene Symbol: Traf3ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044248.1 | NP_001037713.1 | 4, 5 | 1285 | ||
| XM_006256532.2 | XP_006256594.1 | 4, 5 | 1502 | |||
| XM_006256533.2 | XP_006256595.1 | 3, 4 | 997 | |||
| GenBank mRNA | BC100099.1 | - | 4, 5 | 1285 | 7376204 | |

Traf3ip2
Genomic Map