siRNA Details
| Gene Symbol: Cxadrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199406.2 | NP_955438.1 | 3 | 405 | ||
| XM_008766539.1 | XP_008764761.1 | 3 | 1192 | |||
| XM_008766540.1 | XP_008764762.1 | 2 | 512 | |||
| GenBank mRNA | BC063181.1 | - | 3 | 410 | 6887452 | |
| Gene Symbol: LOC103690070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766579.1 | XP_008764801.1 | 1192 | |||
| XM_008766580.1 | XP_008764802.1 | 1192 | ||||
| XM_008766581.1 | XP_008764803.1 | 512 | ||||

Cxadrl1
Genomic Map