siRNA Details
| Gene Symbol: Ssbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760646.1 | XP_008758868.1 | 13 | 1051 | ||
| XM_008760647.1 | XP_008758869.1 | 13 | 1048 | |||
| XM_008760648.1 | XP_008758870.1 | 13 | 1026 | |||
| XM_008760649.1 | XP_008758871.1 | 13 | 1023 | |||
| XM_008760650.1 | XP_008758872.1 | 12 | 961 | |||
| XM_008760651.1 | XP_008758873.1 | 12 | 957 | |||
| XM_008760652.1 | XP_008758874.1 | 12 | 936 | |||
| XM_008760653.1 | XP_008758875.1 | 12 | 932 | |||
| GenBank mRNA | FQ221531.1 | - | 13 | 923 | ||

Ssbp2
Genomic Map