siRNA Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108541.1 | NP_001102011.1 | 8, 9 | 705 | ||
| XM_006231815.2 | XP_006231877.1 | 8, 9 | 766 | |||
| GenBank mRNA | BC166792.1 | - | 8, 9 | 692 | 7937195 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr41| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108541.1 | NP_001102011.1 | 8, 9 | 705 | ||
| XM_006231815.2 | XP_006231877.1 | 8, 9 | 766 | |||
| GenBank mRNA | BC166792.1 | - | 8, 9 | 692 | 7937195 | |