siRNA Details
| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760731.1 | XP_008758953.1 | 2 | 241 | ||
| XM_342191.5 | XP_342192.1 | 2 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufaf2| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760731.1 | XP_008758953.1 | 2 | 241 | ||
| XM_342191.5 | XP_342192.1 | 2 | 239 | |||