siRNA Details
| Gene Symbol: Ankrd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270039.1 | NP_001256968.1 | 1 | 930 | ||
| XM_006231940.2 | XP_006232002.1 | 2 | 662 | |||
| XM_008760737.1 | XP_008758959.1 | 3 | 699 | |||
| XM_008760738.1 | XP_008758960.1 | 2 | 668 | |||
| GenBank mRNA | JQ013731.1 | - | 1 | 930 | ||

Ankrd55
Genomic Map