siRNA Details
| Gene Symbol: Fbxl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108545.1 | NP_001102015.1 | 3 | 1132 | ||
| XM_006232087.2 | XP_006232149.1 | 2 | 3155 | |||
| XM_008760775.1 | XP_008758997.1 | 3 | 924 | |||
| XM_008760776.1 | XP_008758998.1 | 4 | 828 | |||

Fbxl7
Genomic Map