siRNA Details
| Gene Symbol: LOC361914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017465.1 | NP_001017465.1 | 1 | 412 | ||
| XM_006232112.2 | XP_006232174.1 | 1 | 439 | |||
| GenBank mRNA | BC079349.1 | - | 1 | 412 | 7107785 | |
| Gene Symbol: LOC100910566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232118.2 | XP_006232180.1 | 1 | 439 | ||

LOC361914
Genomic Map