siRNA Details
| Gene Symbol: LOC361914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017465.1 | NP_001017465.1 | 4 | 1440 | ||
| XM_006232112.2 | XP_006232174.1 | 5 | 1374 | |||
| GenBank mRNA | BC079349.1 | - | 4 | 1440 | 7107785 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC361914| Gene Symbol: LOC361914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017465.1 | NP_001017465.1 | 4 | 1440 | ||
| XM_006232112.2 | XP_006232174.1 | 5 | 1374 | |||
| GenBank mRNA | BC079349.1 | - | 4 | 1440 | 7107785 | |