siRNA Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017466.2 | NP_001017466.2 | 2 | 372 | ||
| GenBank mRNA | BC092576.1 | - | 2 | 372 | 7122321 | |
| CO383495.1 | - | 2 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp4c| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017466.2 | NP_001017466.2 | 2 | 372 | ||
| GenBank mRNA | BC092576.1 | - | 2 | 372 | 7122321 | |
| CO383495.1 | - | 2 | 350 | |||