siRNA Details
Gene Symbol: Chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017466.2 | NP_001017466.2 | 2 | 423 | ||
GenBank mRNA | BC092576.1 | - | 2 | 423 | 7122321 | |
CO383495.1 | - | 2 | 401 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017466.2 | NP_001017466.2 | 2 | 423 | ||
GenBank mRNA | BC092576.1 | - | 2 | 423 | 7122321 | |
CO383495.1 | - | 2 | 401 |