siRNA Details
| Gene Symbol: Zfand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342213.6 | XP_342214.4 | 7 | 524 | ||
| GenBank mRNA | FQ229099.1 | - | 7 | 510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfand1| Gene Symbol: Zfand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342213.6 | XP_342214.4 | 7 | 524 | ||
| GenBank mRNA | FQ229099.1 | - | 7 | 510 | ||