siRNA Details
| Gene Symbol: Mynn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012178.1 | NP_001012178.1 | 3 | 406 | ||
| XM_006232232.2 | XP_006232294.1 | 3 | 1450 | |||
| XM_006232233.2 | XP_006232295.1 | 3 | 1451 | |||
| XM_006232234.2 | XP_006232296.1 | 3 | 1320 | |||
| GenBank mRNA | BC089853.1 | - | 3 | 406 | 7379696 | |

Mynn
Genomic Map