siRNA Details
Gene Symbol: LOC100366044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760951.1 | XP_008759173.1 | 1 | 141 | ||
XM_008760952.1 | XP_008759174.1 | 1 | 141 |
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083807.1 | - | 144 | 7109902 |
