siRNA Details
| Gene Symbol: RGD1307100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232275.2 | XP_006232337.1 | 24 | 3202 | ||
| XM_008760914.1 | XP_008759136.1 | 24 | 3202 | |||
| XM_008760915.1 | XP_008759137.1 | 24 | 3199 | |||
| XM_008760916.1 | XP_008759138.1 | 24 | 3202 | |||
| XM_008760917.1 | XP_008759139.1 | 24 | 3202 | |||
| XM_008760918.1 | XP_008759140.1 | 23 | 3064 | |||
| XM_008760919.1 | XP_008759141.1 | 24 | 3201 | |||

RGD1307100
Genomic Map