siRNA Details
| Gene Symbol: Spata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108549.1 | NP_001102019.1 | 5 | 1406 | ||
| XM_008760932.1 | XP_008759154.1 | 5 | 1397 | |||
| XM_008760933.1 | XP_008759155.1 | 5 | 1056 | |||
| XM_008760934.1 | XP_008759156.1 | 5 | 1397 | |||

Spata5
Genomic Map