siRNA Details
| Gene Symbol: Intu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760977.1 | XP_008759199.1 | 1536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Intu| Gene Symbol: Intu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760977.1 | XP_008759199.1 | 1536 | |||