siRNA Details
| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008374.1 | NP_001008375.1 | 2, 3 | 555 | ||
| GenBank mRNA | BC083571.1 | - | 2, 3 | 555 | 7112203 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027558, AK014543, AX775753, BC044759, AK184727.

Pgrmc2
Genomic Map