siRNA Details
| Gene Symbol: Pcdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761025.1 | XP_008759247.1 | 2 | 2390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdh10| Gene Symbol: Pcdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761025.1 | XP_008759247.1 | 2 | 2390 | ||