siRNA Details
| Gene Symbol: Elf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012181.2 | NP_001012181.1 | 7 | 2192 | ||
| NM_001033909.1 | NP_001029081.1 | 9 | 1872 | |||
| XM_006232325.2 | XP_006232387.1 | 10 | 2074 | |||
| XM_006232327.2 | XP_006232389.1 | 7 | 4260 | |||
| XM_006232328.1 | XP_006232390.1 | 8 | 1586 | |||
| XM_006232329.1 | XP_006232391.1 | 8 | 1550 | |||
| XM_008760992.1 | XP_008759214.1 | 9 | 2457 | |||
| GenBank mRNA | BC083585.1 | - | 10 | 2024 | 7131145 | |
| BC083598.1 | - | 7 | 1544 | 7132941 | ||

Elf2
Genomic Map