siRNA Details
| Gene Symbol: Veph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014171.1 | NP_001014193.1 | 8 | 1508 | ||
| XM_008761050.1 | XP_008759272.1 | 7 | 1471 | |||
| XM_008761051.1 | XP_008759273.1 | 8 | 1508 | |||
| GenBank mRNA | BC087055.1 | - | 8 | 1508 | 7123921 | |

Veph1
Genomic Map