siRNA Details
| Gene Symbol: Rsrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014172.1 | NP_001014194.1 | 9 | 1033 | ||
| XM_008761055.1 | XP_008759277.1 | 9 | 1023 | |||
| GenBank mRNA | BC087665.1 | - | 9 | 1033 | 7317835 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsrc1| Gene Symbol: Rsrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014172.1 | NP_001014194.1 | 9 | 1033 | ||
| XM_008761055.1 | XP_008759277.1 | 9 | 1023 | |||
| GenBank mRNA | BC087665.1 | - | 9 | 1033 | 7317835 | |