siRNA Details
| Gene Symbol: Kpna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014793.1 | NP_001014793.1 | 9 | 586 | ||
| GenBank mRNA | AY779028.1 | - | 9 | 586 | ||
| FQ224515.1 | - | 587 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kpna4| Gene Symbol: Kpna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014793.1 | NP_001014793.1 | 9 | 586 | ||
| GenBank mRNA | AY779028.1 | - | 9 | 586 | ||
| FQ224515.1 | - | 587 | ||||