siRNA Details
| Gene Symbol: Ppid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004279.1 | NP_001004279.1 | 6, 7 | 820 | ||
| GenBank mRNA | BC076386.1 | - | 6, 7 | 820 | 7108860 | |
| FQ214826.1 | - | 6, 7 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppid| Gene Symbol: Ppid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004279.1 | NP_001004279.1 | 6, 7 | 820 | ||
| GenBank mRNA | BC076386.1 | - | 6, 7 | 820 | 7108860 | |
| FQ214826.1 | - | 6, 7 | 823 | |||