siRNA Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108551.1 | NP_001102021.1 | 8 | 992 | ||
XM_006232517.2 | XP_006232579.1 | 8 | 993 | |||
GenBank mRNA | FQ211169.1 | - | 6 | 546 | ||
FQ223804.1 | - | 6 | 546 |
