siRNA Details
| Gene Symbol: Tmem154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108553.1 | NP_001102023.1 | 2 | 314 | ||
| XM_006232538.2 | XP_006232600.1 | 2 | 489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem154| Gene Symbol: Tmem154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108553.1 | NP_001102023.1 | 2 | 314 | ||
| XM_006232538.2 | XP_006232600.1 | 2 | 489 | |||