siRNA Details
| Gene Symbol: Rnf115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108560.1 | NP_001102030.1 | 4 | 412 | ||
| XM_006232975.1 | XP_006233037.1 | 4 | 433 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf115| Gene Symbol: Rnf115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108560.1 | NP_001102030.1 | 4 | 412 | ||
| XM_006232975.1 | XP_006233037.1 | 4 | 433 | |||