siRNA Details
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013201.1 | NP_001013219.1 | 8 | 1087 | ||
| XM_006233032.2 | XP_006233094.1 | 9 | 1185 | |||
| XM_006233033.2 | XP_006233095.1 | 8 | 1135 | |||
| GenBank mRNA | BC082000.1 | - | 8 | 1087 | 7131173 | |
| FQ211326.1 | - | 10 | 1268 | |||

Gdap2
Genomic Map