siRNA Details
| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277236.1 | NP_001264165.1 | 3 | 587 | ||
| GenBank mRNA | FM044099.1 | - | 3 | 587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb561d1| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277236.1 | NP_001264165.1 | 3 | 587 | ||
| GenBank mRNA | FM044099.1 | - | 3 | 587 | ||