siRNA Details
| Gene Symbol: RGD1309139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134578.1 | NP_001128050.1 | 5 | 814 | ||
| XM_008761408.1 | XP_008759630.1 | 4 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309139| Gene Symbol: RGD1309139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134578.1 | NP_001128050.1 | 5 | 814 | ||
| XM_008761408.1 | XP_008759630.1 | 4 | 800 | |||