siRNA Details
Gene Symbol: RGD1309139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134578.1 | NP_001128050.1 | 6 | 867 | ||
XM_008761408.1 | XP_008759630.1 | 5 | 853 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1309139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134578.1 | NP_001128050.1 | 6 | 867 | ||
XM_008761408.1 | XP_008759630.1 | 5 | 853 |