siRNA Details
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191962.1 | NP_001178891.1 | 14 | 1826 | ||
XM_008761409.1 | XP_008759631.1 | 15 | 2353 | |||
XM_008761410.1 | XP_008759632.1 | 15 | 2140 | |||
XM_008761411.1 | XP_008759633.1 | 14 | 2049 |
