siRNA Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191962.1 | NP_001178891.1 | 7, 8 | 947 | ||
| XM_008761409.1 | XP_008759631.1 | 8, 9 | 1474 | |||
| XM_008761410.1 | XP_008759632.1 | 8, 9 | 1261 | |||
| XM_008761411.1 | XP_008759633.1 | 7, 8 | 1170 | |||

Gpsm2
Genomic Map