siRNA Details
| Gene Symbol: Agl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108564.1 | NP_001102034.1 | 19, 20 | 3097 | ||
| XM_006233229.2 | XP_006233291.1 | 19, 20 | 2738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agl| Gene Symbol: Agl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108564.1 | NP_001102034.1 | 19, 20 | 3097 | ||
| XM_006233229.2 | XP_006233291.1 | 19, 20 | 2738 | |||