siRNA Details
| Gene Symbol: Ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108566.1 | NP_001102036.1 | 2, 3 | 277 | ||
| GenBank mRNA | BC159425.1 | - | 2, 3 | 263 | 9030828 | |
| FQ219812.1 | - | 2, 3 | 283 | |||
| FQ219920.1 | - | 2, 3 | 291 | |||
| FQ220255.1 | - | 2, 3 | 259 | |||
| FQ230162.1 | - | 2, 3 | 259 | |||
| FQ234814.1 | - | 2, 3 | 239 | |||

Ostc
Genomic Map