siRNA Details
| Gene Symbol: Rpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108567.2 | NP_001102037.1 | 5 | 317 | ||
| XM_006233314.2 | XP_006233376.1 | 5 | 482 | |||
| XM_006233315.1 | XP_006233377.1 | 4 | 305 | |||
| GenBank mRNA | BC167113.1 | - | 5 | 317 | 6889959 | |
| FQ211292.1 | - | 5 | 320 | |||
| FQ211369.1 | - | 5 | 319 | |||
| FQ216454.1 | - | 678 | ||||
| FQ221198.1 | - | 5 | 319 | |||
| FQ221629.1 | - | 5 | 319 | |||
| FQ221933.1 | - | 5 | 319 | |||
| FQ223193.1 | - | 5 | 319 | |||
| FQ224761.1 | - | 5 | 325 | |||
| FQ229743.1 | - | 5 | 319 | |||

Rpl34
Genomic Map