siRNA Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004090.2 | NP_001004090.2 | 5 | 494 | ||
| GenBank mRNA | AF540877.1 | - | 5 | 520 | ||
| BC100135.1 | - | 5 | 494 | 7315329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan5| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004090.2 | NP_001004090.2 | 5 | 494 | ||
| GenBank mRNA | AF540877.1 | - | 5 | 520 | ||
| BC100135.1 | - | 5 | 494 | 7315329 | ||