siRNA Details
Gene Symbol: Clca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013202.2 | NP_001013220.1 | 7 | 998 | ||
GenBank mRNA | AB119249.1 | - | 7 | 999 | ||
AB212889.1 | - | 7 | 999 | |||
AB306511.1 | - | 7 | 999 |
