siRNA Details
| Gene Symbol: Fbxw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025730.2 | NP_001020901.1 | 6 | 836 | ||
| XM_006233623.2 | XP_006233685.1 | 5 | 1195 | |||
| XM_006233624.2 | XP_006233686.1 | 7 | 898 | |||
| XM_006233625.2 | XP_006233687.1 | 7 | 1140 | |||
| XM_006233626.2 | XP_006233688.1 | 6 | 792 | |||
| XM_006233627.2 | XP_006233689.1 | 6 | 1036 | |||
| XM_006233628.2 | XP_006233690.1 | 5 | 552 | |||
| GenBank mRNA | BC099179.1 | - | 6 | 839 | 7383052 | |

Fbxw5
Genomic Map