siRNA Details
| Gene Symbol: CD84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184879.1 | NP_001171808.1 | 3 | 648 | ||
| NM_001184881.1 | NP_001171810.1 | 3 | 648 | |||
| NM_001184882.1 | NP_001171811.1 | 2 | 306 | |||
| NM_003874.3 | NP_003865.1 | 3 | 648 | |||
| XM_011510094.2 | XP_011508396.1 | 3 | 629 | |||
| XM_011510095.2 | XP_011508397.1 | 2 | 305 | |||
| GenBank mRNA | AF054815.1 | - | 3 | 638 | ||
| AF054816.1 | - | 3 | 638 | |||
| AF054817.1 | - | 3 | 638 | |||
| AF054818.1 | - | 3 | 638 | |||
| AJ223324.1 | - | 3 | 613 | |||
| AK296290.1 | - | 2 | 267 | |||
| AK313496.1 | - | 3 | 614 | |||
| BC020063.1 | - | 3 | 593 | 4385965 | ||
| CR541847.1 | - | 3 | 569 | |||
| CR933666.1 | - | 2 | 267 | |||
| U82988.1 | - | 3 | 610 | |||
| U96627.1 | - | 3 | 638 | |||
| Y12632.1 | - | 3 | 638 | |||

CD84
Genomic Map