siRNA Details
| Gene Symbol: Mrps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108576.1 | NP_001102046.1 | 4 | 364 | ||
| XM_006233832.2 | XP_006233894.1 | 3 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps2| Gene Symbol: Mrps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108576.1 | NP_001102046.1 | 4 | 364 | ||
| XM_006233832.2 | XP_006233894.1 | 3 | 356 | |||