siRNA Details
| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079941.1 | NP_001073410.1 | 10 | 1512 | ||
| XM_006233835.2 | XP_006233897.1 | 10 | 1521 | |||
| GenBank mRNA | BC129109.1 | - | 10 | 1512 | 7105121 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf3c5| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079941.1 | NP_001073410.1 | 10 | 1512 | ||
| XM_006233835.2 | XP_006233897.1 | 10 | 1521 | |||
| GenBank mRNA | BC129109.1 | - | 10 | 1512 | 7105121 | |