siRNA Details
| Gene Symbol: Setx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233886.2 | XP_006233948.1 | 8 | 1223 | ||
| XM_006233887.2 | XP_006233949.1 | 9 | 1226 | |||
| XM_006233888.2 | XP_006233950.1 | 8 | 1222 | |||
| XM_008761681.1 | XP_008759903.1 | 8 | 1337 | |||
| XM_008761682.1 | XP_008759904.1 | 5 | 591 | |||

Setx
Genomic Map