siRNA Details
| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034938.1 | NP_001030110.1 | 2 | 550 | ||
| GenBank mRNA | AB221075.1 | - | 2 | 550 | ||
| FQ232745.1 | - | 2 | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Endog| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034938.1 | NP_001030110.1 | 2 | 550 | ||
| GenBank mRNA | AB221075.1 | - | 2 | 550 | ||
| FQ232745.1 | - | 2 | 600 | |||