siRNA Details
| Gene Symbol: Sh2d3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108579.1 | NP_001102049.1 | 7 | 1651 | ||
| XM_008761759.1 | XP_008759981.1 | 6 | 2518 | |||
| XM_008761760.1 | XP_008759982.1 | 5 | 2527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d3c| Gene Symbol: Sh2d3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108579.1 | NP_001102049.1 | 7 | 1651 | ||
| XM_008761759.1 | XP_008759981.1 | 6 | 2518 | |||
| XM_008761760.1 | XP_008759982.1 | 5 | 2527 | |||