siRNA Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007744.1 | NP_001007745.1 | 1 | 142 | ||
| GenBank mRNA | BC078943.1 | - | 1 | 142 | 7109352 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor2a| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007744.1 | NP_001007745.1 | 1 | 142 | ||
| GenBank mRNA | BC078943.1 | - | 1 | 142 | 7109352 | |