siRNA Details
| Gene Symbol: Fam129b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109885.1 | NP_001103355.1 | 2 | 288 | ||
| GenBank mRNA | BC168248.1 | - | 2 | 210 | 5598117 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam129b| Gene Symbol: Fam129b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109885.1 | NP_001103355.1 | 2 | 288 | ||
| GenBank mRNA | BC168248.1 | - | 2 | 210 | 5598117 | |