siRNA Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108581.1 | NP_001102051.1 | 14 | 3277 | ||
| XM_006234155.2 | XP_006234217.1 | 13 | 3059 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epc2| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108581.1 | NP_001102051.1 | 14 | 3277 | ||
| XM_006234155.2 | XP_006234217.1 | 13 | 3059 | |||