siRNA Details
| Gene Symbol: Lypd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134580.1 | NP_001128052.1 | 9 | 790 | ||
| XM_008761762.1 | XP_008759984.1 | 6 | 2636 | |||
| XM_008761763.1 | XP_008759985.1 | 7 | 3286 | |||
| XM_008761764.1 | XP_008759986.1 | 6 | 3102 | |||
| XM_008761765.1 | XP_008759987.1 | 8 | 3406 | |||
| XM_008761766.1 | XP_008759988.1 | 5 | 2576 | |||
| XM_008761767.1 | XP_008759989.1 | 5 | 3042 | |||
| XM_008761768.1 | XP_008759990.1 | 5 | 2086 | |||
| XM_008761769.1 | XP_008759991.1 | 6 | 2542 | |||

Lypd6b
Genomic Map